COM448684791 Combi-Blocks, Inc.

2-(4-Hydroxyphenyl)benzoic acid

  • MDL number MFCD04117368
  • Cas Number : 67526-82-3
  • Molecular Weight : 214.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 98

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?YA-0991
Availability details for
Quantity
  • Availability
  • Add to Cart