COM448684805 Combi-Blocks, Inc.

2-(3,5-Dimethylphenyl)benzoic acid

  • MDL number MFCD12859171
  • Cas Number : 1183804-03-6
  • Molecular Weight : 226.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1033

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1033
Availability details for
Quantity
  • Availability
  • Add to Cart