COM448684812 Combi-Blocks, Inc.

2-(3-Methoxyphenyl)benzoic acid

  • MDL number MFCD03426465
  • Cas Number : 38087-96-6
  • Molecular Weight : 228.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1048

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1048
Availability details for
Quantity
  • Availability
  • Add to Cart