COM448684814 Combi-Blocks, Inc.

2-(4-Methoxyphenyl)benzoic acid

  • MDL number MFCD03426469
  • Cas Number : 18110-71-9
  • Molecular Weight : 228.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4'-methoxy[1,1'-biphenyl]-2-carboxylic acid
Inchi Code 1S/C14H12O3/c1-17-11-8-6-10(7-9-11)12-4-2-3-5-13(12)14(15)16/h2-9H,1H3,(H,15,16)
Inchi Key IPLPMLYQQDEHHD-UHFFFAOYSA-N

Description

Purity 96%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1051

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1051
Availability details for
Quantity
  • Availability
  • Add to Cart