COM448684817 Combi-Blocks, Inc.

3-(3-Fluoro-4-methylphenyl)benzoic acid

  • MDL number MFCD06858728
  • Cas Number : 885964-56-7
  • Molecular Weight : 230.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1064

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1064
Availability details for
Quantity
  • Availability
  • Add to Cart