COM448684836 Combi-Blocks, Inc.

2-(3,5-Difluorophenyl)benzoic acid

  • MDL number MFCD03839969
  • Cas Number : 886363-21-9
  • Molecular Weight : 234.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1105

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1105
Availability details for
Quantity
  • Availability
  • Add to Cart