COM448684840 Combi-Blocks, Inc.

4-(2,6-Difluorophenyl)benzoic acid

  • MDL number MFCD18312610
  • Cas Number : 505082-79-1
  • Molecular Weight : 234.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1113

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1113
Availability details for
Quantity
  • Availability
  • Add to Cart