COM448684843 Combi-Blocks, Inc.

2-(3-Acetylphenyl)benzoic acid

  • MDL number MFCD03839957
  • Cas Number : 408367-33-9
  • Molecular Weight : 240.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1117

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1117
Availability details for
Quantity
  • Availability
  • Add to Cart