COM448684844 Combi-Blocks, Inc.

4-(3-Acetylphenyl)benzoic acid

  • MDL number MFCD03839959
  • Cas Number : 199678-04-1
  • Molecular Weight : 240.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3'-acetyl[1,1'-biphenyl]-4-carboxylic acid
Inchi Code 1S/C15H12O3/c1-10(16)13-3-2-4-14(9-13)11-5-7-12(8-6-11)15(17)18/h2-9H,1H3,(H,17,18)
Inchi Key NNPZXRZBAVBTGL-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1119

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1119
Availability details for
Quantity
  • Availability
  • Add to Cart