COM448684846 Combi-Blocks, Inc.

3-(3-Aminocarbonylphenyl)benzoic acid

  • MDL number MFCD18312618
  • Cas Number : 1261997-49-2
  • Molecular Weight : 241.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1130

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1130
Availability details for
Quantity
  • Availability
  • Add to Cart