COM448684847 Combi-Blocks, Inc.

4-(3-Aminocarbonylphenyl)benzoic acid

  • MDL number MFCD18312619
  • Cas Number : 199678-18-7
  • Molecular Weight : 241.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1131

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1131
Availability details for
Quantity
  • Availability
  • Add to Cart