COM448684848 Combi-Blocks, Inc.

2-(4-Carboxyphenyl)benzoic acid

  • MDL number MFCD06203580
  • Cas Number : 606-80-4
  • Molecular Weight : 242.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1135

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1135
Availability details for
Quantity
  • Availability
  • Add to Cart