COM448684851 Combi-Blocks, Inc.

3-(3,4-Methylenedioxyphenyl)benzoic acid

  • MDL number MFCD03426479
  • Cas Number : 24351-56-2
  • Molecular Weight : 242.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1139

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1139
Availability details for
Quantity
  • Availability
  • Add to Cart