COM448684854 Combi-Blocks, Inc.

3-(4-Methoxy-2-methylphenyl)benzoic acid

  • MDL number MFCD11920068
  • Cas Number : 1178070-12-6
  • Molecular Weight : 242.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1145

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1145
Availability details for
Quantity
  • Availability
  • Add to Cart