COM448684855 Combi-Blocks, Inc.

2-(3-Ethoxyphenyl)benzoic acid

  • MDL number MFCD03426472
  • Cas Number : 669713-70-6
  • Molecular Weight : 242.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1150

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1150
Availability details for
Quantity
  • Availability
  • Add to Cart