COM448684865 Combi-Blocks, Inc.

3-(5-Fluoro-2-methoxyphenyl)benzoic acid

  • MDL number MFCD12859510
  • Cas Number : 376592-43-7
  • Molecular Weight : 246.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1181

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1181
Availability details for
Quantity
  • Availability
  • Add to Cart