COM448684869 Combi-Blocks, Inc.

3-(4-Chloro-2-methylphenyl)benzoic acid

  • MDL number MFCD16657009
  • Cas Number : 916220-05-8
  • Molecular Weight : 246.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1196

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1196
Availability details for
Quantity
  • Availability
  • Add to Cart