COM448684872 Combi-Blocks, Inc.

2-(4-t-Butylphenyl)benzoic acid

  • MDL number MFCD06802636
  • Cas Number : 84392-26-7
  • Molecular Weight : 254.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1222

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1222
Availability details for
Quantity
  • Availability
  • Add to Cart