COM448684885 Combi-Blocks, Inc.

3-(4-Carboxy-3-fluorophenyl)benzoic acid

  • MDL number MFCD18312663
  • Cas Number : 1261915-32-5
  • Molecular Weight : 260.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1262

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1262
Availability details for
Quantity
  • Availability
  • Add to Cart