COM448684889 Combi-Blocks, Inc.

2-(4-Chloro-2-methoxyphenyl)benzoic acid

  • MDL number MFCD18312673
  • Cas Number : 1261896-06-3
  • Molecular Weight : 262.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1279

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1279
Availability details for
Quantity
  • Availability
  • Add to Cart