COM448684890 Combi-Blocks, Inc.

4-(2-Trifluoromethylphenyl)benzoic acid

  • MDL number MFCD03424605
  • Cas Number : 198205-79-7
  • Molecular Weight : 266.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1287

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1287
Availability details for
Quantity
  • Availability
  • Add to Cart