COM448684896 Combi-Blocks, Inc.

4-(2,5-Dichlorophenyl)benzoic acid

  • MDL number MFCD11574808
  • Cas Number : 63028-31-9
  • Molecular Weight : 267.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1302

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1302
Availability details for
Quantity
  • Availability
  • Add to Cart