COM448684897 Combi-Blocks, Inc.

2-(3,4-Dichlorophenyl)benzoic acid

  • MDL number MFCD09042379
  • Cas Number : 916849-01-9
  • Molecular Weight : 267.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1303

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1303
Availability details for
Quantity
  • Availability
  • Add to Cart