COM448684907 Combi-Blocks, Inc.

2-(4-Trifluoromethoxyphenyl)benzoic acid

  • MDL number MFCD04117417
  • Cas Number : 408366-18-7
  • Molecular Weight : 282.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1357

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1357
Availability details for
Quantity
  • Availability
  • Add to Cart