COM448684908 Combi-Blocks, Inc.

4-(4-Trifluoromethoxyphenyl)benzoic acid

  • MDL number MFCD04117415
  • Cas Number : 728919-11-7
  • Molecular Weight : 282.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1359

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1359
Availability details for
Quantity
  • Availability
  • Add to Cart