COM448684909 Combi-Blocks, Inc.

4-(2-Fluoro-3-trifluoromethylphenyl)benzoic acid

  • MDL number MFCD18312719
  • Cas Number : 1261896-07-4
  • Molecular Weight : 284.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1362

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1362
Availability details for
Quantity
  • Availability
  • Add to Cart