COM448684911 Combi-Blocks, Inc.

4-(4-Fluoro-3-trifluoromethylphenyl)benzoic acid

  • MDL number MFCD08457042
  • Cas Number : 942475-07-2
  • Molecular Weight : 284.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1365

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1365
Availability details for
Quantity
  • Availability
  • Add to Cart