COM448684914 Combi-Blocks, Inc.

3-(2-Benzyloxyphenyl)benzoic acid

  • MDL number MFCD06802260
  • Cas Number : 893736-38-4
  • Molecular Weight : 304.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1397

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1397
Availability details for
Quantity
  • Availability
  • Add to Cart