COM448684917 Combi-Blocks, Inc.

4-(3-Benzyloxyphenyl)benzoic acid

  • MDL number MFCD06802256
  • Cas Number : 122294-08-0
  • Molecular Weight : 304.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1401

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1401
Availability details for
Quantity
  • Availability
  • Add to Cart