COM448684919 Combi-Blocks, Inc.

4-(4-Benzyloxyphenyl)benzoic acid

  • MDL number MFCD06802253
  • Cas Number : 111153-16-3
  • Molecular Weight : 304.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1404

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1404
Availability details for
Quantity
  • Availability
  • Add to Cart