COM448684920 Combi-Blocks, Inc.

2-(3-BOC-Aminophenyl)benzoic acid

  • MDL number MFCD09055095
  • Cas Number : 927801-48-7
  • Molecular Weight : 313.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1417

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1417
Availability details for
Quantity
  • Availability
  • Add to Cart