COM448684922 Combi-Blocks, Inc.

2-(4-BOC-Aminophenyl)benzoic acid

  • MDL number MFCD09055097
  • Cas Number : 927801-54-5
  • Molecular Weight : 313.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1420

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1420
Availability details for
Quantity
  • Availability
  • Add to Cart