COM448684929 Combi-Blocks, Inc.

4-(2-Methylphenyl)phenol

  • MDL number MFCD06802303
  • Cas Number : 38262-85-0
  • Molecular Weight : 184.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1455

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1455
Availability details for
Quantity
  • Availability
  • Add to Cart