COM448684935 Combi-Blocks, Inc.

2-(3-Aminophenyl)phenol

  • MDL number MFCD18312779
  • Cas Number : 161887-01-0
  • Molecular Weight : 185.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1462

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1462
Availability details for
Quantity
  • Availability
  • Add to Cart