COM448684936 Combi-Blocks, Inc.

3-(3-Aminophenyl)phenol

  • MDL number MFCD04117380
  • Cas Number : 779341-19-4
  • Molecular Weight : 185.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1463

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1463
Availability details for
Quantity
  • Availability
  • Add to Cart