COM448684939 Combi-Blocks, Inc.

4-(3-Hydroxyphenyl)phenol

  • MDL number MFCD11895234
  • Cas Number : 18855-13-5
  • Molecular Weight : 186.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1470

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1470
Availability details for
Quantity
  • Availability
  • Add to Cart