COM448684941 Combi-Blocks, Inc.

4-(2-Fluorophenyl)phenol

  • MDL number MFCD06802292
  • Cas Number : 321-62-0
  • Molecular Weight : 188.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1476

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1476
Availability details for
Quantity
  • Availability
  • Add to Cart