COM448684943 Combi-Blocks, Inc.

3-(4-Fluorophenyl)phenol

  • MDL number MFCD06802305
  • Cas Number : 10540-41-7
  • Molecular Weight : 188.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1481

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1481
Availability details for
Quantity
  • Availability
  • Add to Cart