COM448684949 Combi-Blocks, Inc.

3-(3-Formylphenyl)phenol

  • MDL number MFCD04117377
  • Cas Number : 400745-17-7
  • Molecular Weight : 198.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1496

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1496
Availability details for
Quantity
  • Availability
  • Add to Cart