COM448684950 Combi-Blocks, Inc.

4-(3-Formylphenyl)phenol

  • MDL number MFCD04117372
  • Cas Number : 283147-95-5
  • Molecular Weight : 198.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1497

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1497
Availability details for
Quantity
  • Availability
  • Add to Cart