COM448684951 Combi-Blocks, Inc.

2-(4-Formylphenyl)phenol

  • MDL number MFCD06801753
  • Cas Number : 400744-38-9
  • Molecular Weight : 198.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1498

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1498
Availability details for
Quantity
  • Availability
  • Add to Cart