COM448684959 Combi-Blocks, Inc.

4-(3-Hydroxymethylphenyl)phenol

  • MDL number MFCD17677632
  • Cas Number : 191724-10-4
  • Molecular Weight : 200.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1521

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1521
Availability details for
Quantity
  • Availability
  • Add to Cart