COM448684960 Combi-Blocks, Inc.

4-(4-Hydroxymethylphenyl)phenol

  • MDL number MFCD17677654
  • Cas Number : 162957-24-6
  • Molecular Weight : 200.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1524

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1524
Availability details for
Quantity
  • Availability
  • Add to Cart