COM448684967 Combi-Blocks, Inc.

4-(2-Fluoro-5-methylphenyl)phenol

  • MDL number MFCD18312801
  • Cas Number : 1261911-67-4
  • Molecular Weight : 202.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1539

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1539
Availability details for
Quantity
  • Availability
  • Add to Cart