COM448684970 Combi-Blocks, Inc.

4-(4-Fluoro-3-methylphenyl)phenol

  • MDL number MFCD18312810
  • Cas Number : 64465-60-7
  • Molecular Weight : 202.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1548

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1548
Availability details for
Quantity
  • Availability
  • Add to Cart