COM448684972 Combi-Blocks, Inc.

4-(5-Formylthiophen-2-yl)phenol

  • MDL number MFCD06802776
  • Cas Number : 893740-97-1
  • Molecular Weight : 204.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1569

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1569
Availability details for
Quantity
  • Availability
  • Add to Cart