COM448684982 Combi-Blocks, Inc.

4-(3,5-Difluorophenyl)phenol

  • MDL number MFCD16484203
  • Cas Number : 656304-67-5
  • Molecular Weight : 206.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1587

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1587
Availability details for
Quantity
  • Availability
  • Add to Cart