COM448684989 Combi-Blocks, Inc.

3-(3-Acetylphenyl)phenol

  • MDL number MFCD11933363
  • Cas Number : 365427-02-7
  • Molecular Weight : 212.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1598

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1598
Availability details for
Quantity
  • Availability
  • Add to Cart