COM448684992 Combi-Blocks, Inc.

4-(3-Aminocarbonylphenyl)phenol

  • MDL number MFCD18312842
  • Cas Number : 477549-30-7
  • Molecular Weight : 213.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1611

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1611
Availability details for
Quantity
  • Availability
  • Add to Cart