COM448684995 Combi-Blocks, Inc.

3-(3-Nitrophenyl)phenol

  • MDL number MFCD09909461
  • Cas Number : 136539-67-8
  • Molecular Weight : 215.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1634

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1634
Availability details for
Quantity
  • Availability
  • Add to Cart